siRNA Details
| Gene Symbol: BBS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018190.3 | NP_060660.2 | 4 | 459 | ||
| NM_176824.2 | NP_789794.1 | 4 | 459 | |||
| XM_005263106.3 | XP_005263163.1 | 4 | 482 | |||
| XM_011532079.2 | XP_011530381.1 | 4 | 482 | |||
| XM_011532080.2 | XP_011530382.1 | 4 | 482 | |||
| XM_011532081.2 | XP_011530383.1 | 4 | 482 | |||
| XM_017008357.1 | XP_016863846.1 | 4 | 482 | |||
| XM_017008358.1 | XP_016863847.1 | 4 | 482 | |||
| GenBank mRNA | AF521643.1 | - | 4 | 423 | ||
| AF521644.1 | - | 4 | 423 | |||
| AK001577.1 | - | 4 | 423 | |||
| AK308076.1 | - | 4 | 450 | |||
| AK309803.1 | - | 3 | 397 | |||
| BC032691.1 | - | 4 | 376 | 5240398 | ||
| BG720664.1 | - | 4 | 455 | |||
| BQ940332.1 | - | 4 | 424 | |||
| BX510161.1 | - | 4 | 460 | |||

BBS7
Genomic Map