siRNA Details
| Gene Symbol: CRYM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026675.1 | 3 | 653 | |||
| GenBank mRNA | AK123760.1 | - | 3 | 654 | ||
| BC056679.1 | - | 3 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYM-AS1| Gene Symbol: CRYM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026675.1 | 3 | 653 | |||
| GenBank mRNA | AK123760.1 | - | 3 | 654 | ||
| BC056679.1 | - | 3 | 381 | |||