siRNA Details
| Gene Symbol: FAM136A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032822.2 | NP_116211.2 | 2 | 279 | ||
| XM_005264620.1 | XP_005264677.1 | 3 | 329 | |||
| GenBank mRNA | AK027574.1 | - | 2 | 260 | ||
| AK225148.1 | - | 2 | 216 | |||
| AK225151.1 | - | 2 | 247 | |||
| AK309543.1 | - | 2 | 870 | |||
| AK315877.1 | - | 3 | 443 | |||
| AL833147.1 | - | 2 | 222 | |||
| BC014975.1 | - | 2 | 237 | 4903550 | ||
| DB266394.1 | - | 2 | 281 | |||

FAM136A
Genomic Map