siRNA Details
| Gene Symbol: RP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178857.5 | NP_849188.4 | 4 | 4197 | ||
| GenBank mRNA | AJ491324.2 | - | 4244 | |||
| AY168341.1 | - | 4 | 4198 | |||
| AY168342.1 | - | 4 | 4246 | |||
| AY168343.1 | - | 4 | 4246 | |||
| AY168344.1 | - | 4246 | ||||
| AY168345.1 | - | 4246 | ||||
| AY168346.1 | - | 4198 | ||||

RP1L1
Genomic Map