siRNA Details
| Gene Symbol: TTC21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024753.4 | NP_079029.3 | 10, 11 | 1316 | ||
| XM_006712761.1 | XP_006712824.1 | 10, 11 | 1259 | |||
| XM_011511871.2 | XP_011510173.1 | 5, 6 | 4561 | |||
| XM_011511872.2 | XP_011510174.1 | 10, 11 | 1259 | |||
| XM_017004967.1 | XP_016860456.1 | 10, 11 | 1259 | |||
| XM_017004968.1 | XP_016860457.1 | 5, 6 | 2807 | |||
| XM_017004969.1 | XP_016860458.1 | 8, 9 | 976 | |||
| GenBank mRNA | AB073395.1 | - | 10, 11 | 1223 | ||
| AK057268.1 | - | 10, 11 | 1254 | |||
| AK292942.1 | - | 10, 11 | 1259 | |||
| BC055424.1 | - | 10, 11 | 1316 | 4837489 | ||

TTC21B
Genomic Map