siRNA Details
| Gene Symbol: TTC21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024753.4 | NP_079029.3 | 14 | 1880 | ||
| XM_006712761.1 | XP_006712824.1 | 14 | 1823 | |||
| XM_011511871.2 | XP_011510173.1 | 9 | 5125 | |||
| XM_011511872.2 | XP_011510174.1 | 14 | 1823 | |||
| XM_017004967.1 | XP_016860456.1 | 14 | 1823 | |||
| XM_017004968.1 | XP_016860457.1 | 9 | 3371 | |||
| XM_017004969.1 | XP_016860458.1 | 12 | 1540 | |||
| GenBank mRNA | AK057268.1 | - | 14 | 1818 | ||
| AK292942.1 | - | 14 | 1823 | |||
| BC055424.1 | - | 14 | 1880 | 4837489 | ||

TTC21B
Genomic Map