siRNA Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276503.1 | NP_001263432.1 | 3 | 1166 | ||
| NM_001276504.1 | NP_001263433.1 | 3 | 1194 | |||
| NM_001276506.1 | NP_001263435.1 | 5 | 1441 | |||
| NM_003002.3 | NP_002993.1 | 4 | 1311 | |||
| NR_077060.1 | 5 | 1365 | ||||
| GenBank mRNA | AB006202.1 | - | 4 | 1238 | ||
| AK075360.1 | - | 4 | 1237 | |||
| BC005263.1 | - | 4 | 1255 | 3930691 | ||
| BC009574.1 | - | 4 | 1236 | 4063808 | ||
| BC012603.1 | - | 4 | 1240 | 4108104 | ||
| BC015188.1 | - | 4 | 1262 | 4279214 | ||
| BC015992.1 | - | 4 | 1258 | 4292983 | ||
| BC022350.1 | - | 4 | 1250 | 4275516 | ||
| BC070307.1 | - | 4 | 1236 | 4277540 | ||
| BC071755.1 | - | 4 | 1254 | 4774525 | ||
| BC071756.1 | - | 4 | 1236 | 6450992 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024382.1 | - | 1 | 124 | Chr. 1 - 25289838 - 25293540 [-] on Build GRCh38 | |

SDHD
Genomic Map