siRNA Details
| Gene Symbol: SOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018013.3 | NP_060483.3 | 4 | 942 | ||
| XM_005267041.4 | XP_005267098.1 | 5 | 1103 | |||
| XM_005267042.4 | XP_005267099.1 | 5 | 1104 | |||
| XM_011535920.2 | XP_011534222.1 | 5 | 1103 | |||
| XM_011535921.2 | XP_011534223.1 | 5 | 627 | |||
| XM_017010991.1 | XP_016866480.1 | 3 | 1465 | |||
| XM_017010992.1 | XP_016866481.1 | 3 | 234 | |||
| XM_017010993.1 | XP_016866482.1 | 3 | 264 | |||
| XM_017010994.1 | XP_016866483.1 | 3 | 223 | |||
| GenBank mRNA | AK090879.1 | - | 4 | 546 | ||
| DQ507800.1 | - | 4 | 438 | |||

SOBP
Genomic Map