siRNA Details
| Gene Symbol: CCDC88C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080414.3 | NP_001073883.2 | 11 | 1218 | ||
| XM_005267691.4 | XP_005267748.1 | 11 | 1872 | |||
| XM_011536796.2 | XP_011535098.1 | 11 | 1172 | |||
| XM_017021335.1 | XP_016876824.1 | 11 | 1872 | |||
| XM_017021337.1 | XP_016876826.1 | 11 | 1872 | |||
| GenBank mRNA | CR747498.1 | - | 4 | 389 | ||
| CR998718.1 | - | 4 | 318 | |||

CCDC88C
Genomic Map