siRNA Details
| Gene Symbol: CCDC88C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080414.3 | NP_001073883.2 | 26, 27 | 4785 | ||
| XM_011536796.2 | XP_011535098.1 | 26, 27 | 4739 | |||
| XM_017021335.1 | XP_016876824.1 | 26, 27 | 5439 | |||
| XM_017021336.1 | XP_016876825.1 | 11, 12 | 1848 | |||
| GenBank mRNA | BC035914.1 | - | 2, 3 | 497 | ||
| BX248302.1 | - | 1, 2 | 296 | |||

CCDC88C
Genomic Map