siRNA Details
| Gene Symbol: LINC01567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122072.1 | 3 | 1204 | |||
| GenBank mRNA | AK127191.1 | - | 3 | 1204 | ||
| BC132761.1 | - | 1 | 413 | |||
| BC132789.1 | - | 1 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01567| Gene Symbol: LINC01567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122072.1 | 3 | 1204 | |||
| GenBank mRNA | AK127191.1 | - | 3 | 1204 | ||
| BC132761.1 | - | 1 | 413 | |||
| BC132789.1 | - | 1 | 413 | |||