siRNA Details
| Gene Symbol: MYO15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016239.3 | NP_057323.3 | 4 | 4066 | ||
| XM_011523918.2 | XP_011522220.1 | 3 | 4426 | |||
| XM_017024714.1 | XP_016880203.1 | 3 | 4426 | |||
| XM_017024715.1 | XP_016880204.1 | 3 | 4426 | |||
| GenBank mRNA | AB209423.1 | - | 1 | 6036 | ||
| AF144094.1 | - | 4 | 4066 | |||

MYO15A
Genomic Map