siRNA Details
| Gene Symbol: TNPO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191028.2 | NP_001177957.2 | 4 | 908 | ||
| NM_012470.3 | NP_036602.1 | 4 | 908 | |||
| NR_034053.2 | 5 | 1069 | ||||
| XM_011515989.2 | XP_011514291.1 | 4 | 510 | |||
| GenBank mRNA | AF145029.1 | - | 4 | 618 | ||
| AJ133769.1 | - | 4 | 505 | |||
| AK225999.1 | - | 4 | 879 | |||
| AK312580.1 | - | 4 | 635 | |||
| BC068530.1 | - | 4 | 636 | |||

TNPO3
Genomic Map