siRNA Details
| Gene Symbol: TNPO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191028.2 | NP_001177957.2 | 9 | 1635 | ||
| NM_012470.3 | NP_036602.1 | 9 | 1635 | |||
| NR_034053.2 | 10 | 1796 | ||||
| XM_011515989.2 | XP_011514291.1 | 9 | 1237 | |||
| GenBank mRNA | AF145029.1 | - | 9 | 1345 | ||
| AJ133769.1 | - | 9 | 1232 | |||
| AK001263.1 | - | 2 | 189 | |||
| AK225999.1 | - | 9 | 1606 | |||
| AK312580.1 | - | 9 | 1362 | |||
| BC068530.1 | - | 9 | 1363 | |||

TNPO3
Genomic Map