siRNA Details
| Gene Symbol: CYP2U1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183075.2 | NP_898898.1 | 5 | 1668 | ||
| XM_005262717.2 | XP_005262774.1 | 5 | 1734 | |||
| XM_005262720.2 | XP_005262777.1 | 4 | 1044 | |||
| GenBank mRNA | AK290002.1 | - | 5 | 1673 | ||
| AK298989.1 | - | 4 | 590 | |||
| AK303284.1 | - | 6 | 1473 | |||
| AK304724.1 | - | 6 | 1546 | |||
| AY343323.1 | - | 5 | 1624 | |||
| BC132767.1 | - | 5 | 1633 | 40146789 | ||
| BC136483.1 | - | 5 | 1633 | 9020472 | ||

CYP2U1
Genomic Map