siRNA Details
| Gene Symbol: CYP2U1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183075.2 | NP_898898.1 | 2 | 952 | ||
| XM_005262717.2 | XP_005262774.1 | 2 | 1018 | |||
| GenBank mRNA | AK290002.1 | - | 2 | 957 | ||
| AK303284.1 | - | 3 | 757 | |||
| AK304724.1 | - | 3 | 830 | |||
| AY343323.1 | - | 2 | 908 | |||
| BC132767.1 | - | 2 | 917 | 40146789 | ||
| BC136483.1 | - | 2 | 917 | 9020472 | ||
| BX354124.2 | - | 2 | 889 | |||

CYP2U1
Genomic Map