siRNA Details
| Gene Symbol: POGLUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152305.2 | NP_689518.1 | 11 | 3199 | ||
| NR_024265.1 | 11 | 3342 | ||||
| XM_006713705.3 | XP_006713768.1 | 11 | 3151 | |||
| XM_017006878.1 | XP_016862367.1 | 11 | 3314 | |||
| XM_017006879.1 | XP_016862368.1 | 12 | 3460 | |||
| XM_017006880.1 | XP_016862369.1 | 12 | 3317 | |||
| GenBank mRNA | AK027012.1 | - | 2 | 2126 | ||
| AY298903.1 | - | 11 | 3185 | |||

POGLUT1
Genomic Map