siRNA Details
| Gene Symbol: GMPPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013334.3 | NP_037466.2 | 1 | 329 | ||
| NM_021971.2 | NP_068806.1 | 1 | 329 | |||
| GenBank mRNA | AF135421.1 | - | 1 | 221 | ||
| AK024319.1 | - | 1 | 306 | |||
| AK291700.1 | - | 1 | 304 | |||
| BC001141.1 | - | 1 | 268 | 3503869 | ||
| BC008033.1 | - | 1 | 268 | 3355099 | ||
| DA411701.1 | - | 1 | 329 | |||

GMPPB
Genomic Map