siRNA Details
| Gene Symbol: CYP2A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000766.4 | NP_000757.2 | 9 | 1716 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2A13| Gene Symbol: CYP2A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000766.4 | NP_000757.2 | 9 | 1716 | ||