siRNA Details
| Gene Symbol: TULP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289395.1 | NP_001276324.1 | 11 | 1026 | ||
| NM_003322.4 | NP_003313.3 | 12 | 1185 | |||
| GenBank mRNA | AB593124.1 | - | 12 | 1171 | ||
| BC032714.1 | - | 11 | 1011 | 5495136 | ||
| BC065261.1 | - | 11 | 994 | 6144003 | ||
| U82468.1 | - | 12 | 1177 | |||

TULP1
Genomic Map