siRNA Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003673.3 | NP_003664.1 | 2 | 932 | ||
| GenBank mRNA | AJ000491.1 | - | 2 | 928 | ||
| AK096328.1 | - | 2 | 2091 | |||
| BC012628.1 | - | 2 | 933 | 4251080 | ||
| BC013330.1 | - | 2 | 933 | 4273711 | ||

TCAP
Genomic Map