siRNA Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003673.3 | NP_003664.1 | 1 | 89 | ||
| GenBank mRNA | AJ000491.1 | - | 1 | 85 | ||
| AK096328.1 | - | 1 | 1248 | |||
| BC012628.1 | - | 1 | 90 | 4251080 | ||
| BC013330.1 | - | 1 | 90 | 4273711 | ||
| BF670957.1 | - | 1 | 90 | |||
| DA899335.1 | - | 1 | 89 | |||
| KU178429.1 | - | 1 | 75 | |||
| KU178430.1 | - | 1 | 75 | |||

TCAP
Genomic Map