siRNA Details
| Gene Symbol: POMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000939.3 | NP_000930.1 | 3 | 1314 | ||
| NM_001035256.2 | NP_001030333.1 | 4 | 1364 | |||
| NM_001319204.1 | NP_001306133.1 | 4 | 1394 | |||
| NM_001319205.1 | NP_001306134.1 | 3 | 1344 | |||
| GenBank mRNA | AW951218.1 | - | 97 | |||
| BC065832.1 | - | 3 | 1026 | 30392189 | ||
| M28636.1 | - | 1 | 165 | |||
| T29537.1 | - | 1 | 97 | |||

POMC
Genomic Map