siRNA Details
| Gene Symbol: DCANP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130848.2 | NP_570900.1 | 1 | 633 | ||
| GenBank mRNA | AB074498.1 | - | 1 | 633 | ||
| BC069608.1 | - | 1 | 477 | 7262210 | ||
| BC069610.1 | - | 1 | 477 | 7262222 | ||
| BC074831.2 | - | 1 | 419 | 30915544 | ||
| BC074832.2 | - | 1 | 419 | 30915326 | ||

DCANP1
Genomic Map