siRNA Details
| Gene Symbol: MED11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001683.3 | NP_001001683.1 | 3 | 524 | ||
| NM_001305000.1 | NP_001291929.1 | 3 | 635 | |||
| GenBank mRNA | BC070377.1 | - | 3 | 504 | 6450545 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED11| Gene Symbol: MED11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001683.3 | NP_001001683.1 | 3 | 524 | ||
| NM_001305000.1 | NP_001291929.1 | 3 | 635 | |||
| GenBank mRNA | BC070377.1 | - | 3 | 504 | 6450545 | |