siRNA Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184992.1 | NP_001171921.1 | 6 | 1473 | ||
| NM_032322.3 | NP_115698.3 | 5 | 1408 | |||
| NM_197939.1 | NP_922921.1 | 4 | 1245 | |||
| XM_005258043.4 | XP_005258100.1 | 6 | 2920 | |||
| XM_017025223.1 | XP_016880712.1 | 5 | 1594 | |||
| GenBank mRNA | AB470605.1 | - | 5 | 1272 | ||
| AJ496729.1 | - | 5 | 1323 | |||
| AK122646.1 | - | 4 | 1118 | |||
| AK122709.1 | - | 6 | 1203 | |||
| AY598332.1 | - | 5 | 1272 | |||
| BC005084.1 | - | 4 | 1160 | |||
| BC082262.1 | - | 6 | 1351 | |||
| BC126420.1 | - | 5 | 1362 | 8992136 | ||
| BC126422.1 | - | 5 | 1362 | 8992138 | ||
| BC144143.1 | - | 3 | 1055 | |||

RNF135
Genomic Map