siRNA Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 2662 | ||
| XM_005245848.3 | XP_005245905.1 | 7 | 2897 | |||
| XM_005245849.3 | XP_005245906.1 | 7 | 3002 | |||
| XM_005245850.3 | XP_005245907.1 | 8 | 3022 | |||
| XM_005245851.3 | XP_005245908.1 | 7 | 2894 | |||
| XM_005245852.3 | XP_005245909.1 | 6 | 2671 | |||
| XM_011541255.1 | XP_011539557.1 | 7 | 2576 | |||
| XM_011541256.2 | XP_011539558.1 | 7 | 3016 | |||
| XM_011541257.2 | XP_011539559.1 | 6 | 3771 | |||

AHDC1
Genomic Map