siRNA Details
| Gene Symbol: DUOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014080.4 | NP_054799.4 | 18 | 2478 | ||
| XM_005254421.2 | XP_005254478.1 | 18 | 2478 | |||
| GenBank mRNA | AB209010.1 | - | 12 | 4006 | ||
| AF181972.1 | - | 10 | 1261 | |||
| AF230496.1 | - | 18 | 2460 | |||
| AF267981.1 | - | 18 | 2478 | |||

DUOX2
Genomic Map