siRNA Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000133.3 | NP_000124.1 | 8 | 1041 | ||
| NM_001313913.1 | NP_001300842.1 | 7 | 927 | |||
| XM_005262397.4 | XP_005262454.1 | 7 | 928 | |||
| GenBank mRNA | A13997.1 | - | 8 | 1013 | ||
| AB186358.1 | - | 7 | 898 | |||
| AK292749.1 | - | 8 | 1012 | |||
| BC109214.1 | - | 8 | 1041 | 40007636 | ||
| BC109215.1 | - | 8 | 1041 | 40007637 | ||
| FR846238.1 | - | 8 | 1012 | |||
| FR846239.1 | - | 8 | 1012 | |||
| FR846240.1 | - | 8 | 1012 | |||
| J00136.1 | - | 8 | 1015 | |||
| J00137.1 | - | 8 | 1013 | |||
| M11309.1 | - | 8 | 1012 | |||

F9
Genomic Map