siRNA Details
| Gene Symbol: EFR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319099.1 | NP_001306028.1 | 5 | 672 | ||
| NM_014971.1 | NP_055786.1 | 5 | 684 | |||
| XM_011532701.2 | XP_011531003.1 | 5 | 684 | |||
| GenBank mRNA | AB023170.1 | - | 5 | 658 | ||
| AK095190.1 | - | 5 | 672 | |||
| BC049384.1 | - | 2 | 228 | 5742159 | ||

EFR3B
Genomic Map