siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006269.1 | NP_006260.1 | 4 | 4712 | ||
| XM_017013721.1 | XP_016869210.1 | 9 | 5875 | |||
| XM_017013722.1 | XP_016869211.1 | 4 | 4728 | |||
| GenBank mRNA | AF141021.1 | - | 4 | 4712 | ||
| AF146592.1 | - | 4 | 4660 | |||

RP1
Genomic Map