siRNA Details
| Gene Symbol: C17orf102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207454.2 | NP_997337.2 | 2 | 537 | ||
| GenBank mRNA | AK126768.1 | - | 2 | 537 | ||
| BC133004.1 | - | 2 | 468 | 40147026 | ||
| BC136710.1 | - | 2 | 468 | 9020700 | ||
| BC142651.1 | - | 2 | 521 | 40148022 | ||

C17orf102
Genomic Map