siRNA Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174889.4 | NP_777549.1 | 4 | 578 | ||
| GenBank mRNA | AB183433.1 | - | 4 | 451 | ||
| AF087990.1 | - | 4 | 547 | |||
| AK291296.1 | - | 4 | 541 | |||
| BC001753.1 | - | 1 | 500 | 3354845 | ||
| BC033965.1 | - | 4 | 494 | 5288040 | ||
| BC070357.1 | - | 3 | 456 | |||
| KU178834.1 | - | 4 | 451 | |||
| KU178835.1 | - | 3 | 410 | |||
| KU178836.1 | - | 4 | 328 | |||

NDUFAF2
Genomic Map