siRNA Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000349.2 | NP_000340.2 | 6 | 937 | ||
| GenBank mRNA | AF035277.1 | - | 6 | 785 | ||
| AK090513.1 | - | 4 | 1768 | |||
| AK315122.1 | - | 6 | 807 | |||
| AV708514.1 | - | 5 | 371 | |||
| BC010550.1 | - | 6 | 738 | 4153601 | ||
| BX647744.1 | - | 2 | 923 | |||
| CR456803.1 | - | 6 | 673 | |||
| CR541850.1 | - | 6 | 673 | |||
| KU178337.1 | - | 6 | 673 | |||
| S79669.1 | - | 6 | 825 | |||
| U17280.1 | - | 6 | 799 | |||

STAR
Genomic Map