siRNA Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000349.2 | NP_000340.2 | 3 | 461 | ||
| XM_006716392.2 | XP_006716455.1 | 3 | 644 | |||
| GenBank mRNA | AF035277.1 | - | 3 | 309 | ||
| AK090513.1 | - | 2 | 645 | |||
| AK315122.1 | - | 3 | 331 | |||
| AL526126.3 | - | 3 | 468 | |||
| BC010550.1 | - | 3 | 262 | 4153601 | ||
| CR456803.1 | - | 3 | 197 | |||
| CR541850.1 | - | 3 | 197 | |||
| S79669.1 | - | 3 | 349 | |||
| U17280.1 | - | 3 | 323 | |||

STAR
Genomic Map