siRNA Details
| Gene Symbol: TMEM231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077416.2 | NP_001070884.2 | 6 | 2651 | ||
| NM_001077418.2 | NP_001070886.1 | 7 | 2554 | |||
| NR_074083.1 | 7 | 2677 | ||||
| GenBank mRNA | AK025820.1 | - | 7 | 2526 | ||
| AK057689.1 | - | 2 | 2403 | |||
| AK096650.1 | - | 6 | 2603 | |||
| AY358612.1 | - | 7 | 2550 | |||
| BC010609.1 | - | 7 | 2508 | 4214587 | ||
| BC016401.1 | - | 6 | 2186 | |||
| BC063677.1 | - | 7 | 2659 | |||
| Gene Symbol: ABHD14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FJ224306.1 | - | 1509 | |||

TMEM231
Genomic Map