siRNA Details
| Gene Symbol: MCOLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020533.2 | NP_065394.1 | 6, 7 | 925 | ||
| GenBank mRNA | AF249319.1 | - | 6, 7 | 868 | ||
| AF287269.1 | - | 6, 7 | 909 | |||
| AJ293659.1 | - | 5, 6 | 1099 | |||
| AJ293970.1 | - | 6, 7 | 864 | |||
| AK026102.1 | - | 6, 7 | 879 | |||
| AK222673.1 | - | 6, 7 | 904 | |||
| AK294330.1 | - | 6, 7 | 890 | |||
| BC005149.2 | - | 6, 7 | 907 | 3507836 | ||

MCOLN1
Genomic Map