siRNA Details
| Gene Symbol: TMEM127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193304.2 | NP_001180233.1 | 4 | 922 | ||
| NM_017849.3 | NP_060319.1 | 4 | 945 | |||
| XM_017004450.1 | XP_016859939.1 | 3 | 526 | |||
| XM_017004451.1 | XP_016859940.1 | 3 | 571 | |||
| XM_017004452.1 | XP_016859941.1 | 3 | 526 | |||
| GenBank mRNA | AK000514.1 | - | 4 | 926 | ||
| AK293276.1 | - | 6 | 826 | |||
| BC028575.1 | - | 4 | 903 | 4837629 | ||
| BC039892.1 | - | 4 | 898 | 5725347 | ||

TMEM127
Genomic Map