siRNA Details
| Gene Symbol: ANKRD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045839.1 | 10 | 1818 | |||
| GenBank mRNA | AY726574.1 | - | 1 | 1106 | ||
| BC025283.1 | - | 7 | 2583 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD11| Gene Symbol: ANKRD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045839.1 | 10 | 1818 | |||
| GenBank mRNA | AY726574.1 | - | 1 | 1106 | ||
| BC025283.1 | - | 7 | 2583 | |||