siRNA Details
| Gene Symbol: SAMHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015474.3 | NP_056289.2 | 13 | 1653 | ||
| XM_005260384.3 | XP_005260441.1 | 13 | 1626 | |||
| XM_011528761.2 | XP_011527063.1 | 13 | 1626 | |||
| GenBank mRNA | AB013847.1 | - | 13 | 1453 | ||
| AB208944.1 | - | 13 | 1478 | |||
| AF228421.1 | - | 13 | 1606 | |||
| AK027811.1 | - | 13 | 1626 | |||
| AK304187.1 | - | 13 | 1410 | |||
| AK311150.1 | - | 2 | 88 | |||
| AK315169.1 | - | 13 | 1586 | |||
| AL050267.1 | - | 13 | 1528 | |||
| BC036450.2 | - | 13 | 1481 | 4816438 | ||

SAMHD1
Genomic Map