siRNA Details
| Gene Symbol: CNTNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014141.5 | NP_054860.1 | 12, 13 | 2423 | ||
| XM_017011950.1 | XP_016867439.1 | 12, 13 | 2749 | |||
| GenBank mRNA | AB020675.1 | - | 12, 13 | 1932 | ||
| AF319045.1 | - | 12, 13 | 2047 | |||
| AK296532.1 | - | 1, 2 | 79 | |||
| AK315113.1 | - | 12, 13 | 2012 | |||
| BC093780.1 | - | 12, 13 | 1950 | 7939625 | ||
| BC113373.1 | - | 12, 13 | 1950 | 8322425 | ||
| KJ534811.1 | - | 11, 12 | 1796 | |||
| KJ535088.1 | - | 1, 2 | 384 | |||

CNTNAP2
Genomic Map