siRNA Details
Gene Symbol: FLJ26850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027257.1 | 2 | 645 | |||
GenBank mRNA | AK130360.1 | - | 2 | 736 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ26850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027257.1 | 2 | 645 | |||
GenBank mRNA | AK130360.1 | - | 2 | 736 |