siRNA Details
| Gene Symbol: EPG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020964.2 | NP_066015.2 | 12 | 2479 | ||
| XM_011526122.1 | XP_011524424.1 | 12 | 2479 | |||
| XM_017025889.1 | XP_016881378.1 | 12 | 2461 | |||
| XM_017025890.1 | XP_016881379.1 | 12 | 2461 | |||
| XM_017025891.1 | XP_016881380.1 | 12 | 2479 | |||
| XM_017025892.1 | XP_016881381.1 | 11 | 2504 | |||
| XM_017025893.1 | XP_016881382.1 | 5 | 611 | |||
| GenBank mRNA | AB046852.1 | - | 12 | 2414 | ||

EPG5
Genomic Map