siRNA Details
Gene Symbol: GIPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133261.2 | NP_573568.1 | 6 | 2290 | ||
XM_005259492.3 | XP_005259549.1 | 6 | 2343 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GIPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133261.2 | NP_573568.1 | 6 | 2290 | ||
XM_005259492.3 | XP_005259549.1 | 6 | 2343 |