siRNA Details
| Gene Symbol: ZNF772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024596.2 | NP_001019767.1 | 5 | 1175 | ||
| NM_001144068.1 | NP_001137540.1 | 4 | 1052 | |||
| XM_005258943.4 | XP_005259000.1 | 3 | 4305 | |||
| XM_005258944.3 | XP_005259001.1 | 2 | 4266 | |||
| XM_011526980.2 | XP_011525282.1 | 5 | 4809 | |||
| XM_017026813.1 | XP_016882302.1 | 4 | 4686 | |||
| GenBank mRNA | AK294941.1 | - | 3 | 880 | ||
| AK301562.1 | - | 2 | 854 | |||
| AK302574.1 | - | 4 | 1014 | |||
| BX647068.1 | - | 5 | 1175 | |||
| CR749225.1 | - | 2 | 861 | |||

ZNF772
Genomic Map