siRNA Details
| Gene Symbol: INPP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318502.1 | NP_001305431.1 | 8 | 2023 | ||
| NM_019892.5 | NP_063945.2 | 8 | 2026 | |||
| XM_017014926.1 | XP_016870415.1 | 8 | 2065 | |||
| GenBank mRNA | AF187891.1 | - | 8 | 1727 | ||
| AL832846.1 | - | 8 | 2024 | |||
| BC028032.1 | - | 8 | 2008 | 5242186 | ||
| U45974.1 | - | 2 | 240 | |||

INPP5E
Genomic Map