siRNA Details
| Gene Symbol: LZTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006767.3 | NP_006758.2 | 8 | 821 | ||
| GenBank mRNA | AK296347.1 | - | 7 | 702 | ||
| AK310747.1 | - | 6 | 424 | |||
| AK313502.1 | - | 3 | 223 | |||
| BC026214.1 | - | 8 | 723 | 4399716 | ||
| CT841521.1 | - | 8 | 737 | |||
| D38496.1 | - | 8 | 716 | |||

LZTR1
Genomic Map