siRNA Details
| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198859.3 | NP_942559.1 | 6 | 1350 | ||
| XM_011533432.2 | XP_011531734.1 | 7 | 1144 | |||
| XM_011533433.2 | XP_011531735.1 | 7 | 1144 | |||
| XM_011533434.2 | XP_011531736.1 | 7 | 969 | |||
| XM_011533435.2 | XP_011531737.1 | 7 | 969 | |||
| XM_011533436.2 | XP_011531738.1 | 6 | 1202 | |||
| XM_011533437.2 | XP_011531739.1 | 6 | 10272 | |||
| XM_011533438.2 | XP_011531740.1 | 3 | 1094 | |||
| XM_011533440.2 | XP_011531742.1 | 7 | 1144 | |||
| XM_017005798.1 | XP_016861287.1 | 6 | 1074 | |||
| XM_017005799.1 | XP_016861288.1 | 5 | 2823 | |||
| GenBank mRNA | AL833539.1 | - | 6 | 1351 | ||
| BC119002.1 | - | 6 | 1331 | 40082599 | ||
| BX647797.1 | - | 1 | 46 | |||

PRICKLE2
Genomic Map