siRNA Details
| Gene Symbol: ZAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079.3 | NP_001070.2 | 13 | 1911 | ||
| NM_207519.1 | NP_997402.1 | 5 | 929 | |||
| XM_017004867.1 | XP_016860356.1 | 15 | 3312 | |||
| XM_017004868.1 | XP_016860357.1 | 15 | 3294 | |||
| XM_017004869.1 | XP_016860358.1 | 15 | 3312 | |||
| XM_017004870.1 | XP_016860359.1 | 15 | 3312 | |||
| GenBank mRNA | AB083211.1 | - | 5 | 929 | ||
| AL832287.1 | - | 6 | 2756 | |||
| BC039039.1 | - | 12 | 1565 | 5168073 | ||
| BC053878.1 | - | 13 | 1887 | 5757161 | ||
| L05148.1 | - | 13 | 1913 | |||

ZAP70
Genomic Map