siRNA Details
| Gene Symbol: FAM134B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034850.2 | NP_001030022.1 | 9 | 1484 | ||
| NM_019000.4 | NP_061873.2 | 7 | 1360 | |||
| XM_011514053.2 | XP_011512355.1 | 10 | 1543 | |||
| XM_011514054.2 | XP_011512356.1 | 8 | 1494 | |||
| XM_011514055.2 | XP_011512357.1 | 7 | 1284 | |||
| GenBank mRNA | AK000159.1 | - | 1 | 403 | ||
| AK024920.1 | - | 7 | 1190 | |||
| AK025808.1 | - | 9 | 1411 | |||
| AK025832.1 | - | 1 | 406 | |||
| AK225371.1 | - | 1 | 406 | |||
| AL832438.1 | - | 9 | 1160 | |||
| BC030517.1 | - | 3 | 556 | |||
| BC053326.1 | - | 9 | 1408 | 5014868 | ||
| BC073132.1 | - | 7 | 1193 | 5760145 | ||
| CR994722.1 | - | 3 | 599 | |||
| DA136983.1 | - | 1 | 64 | |||

FAM134B
Genomic Map